N-tert-Butylbenzenesulfenamide ≥96.0%

Supplier: TCI America

B2240-25G B2240-5G B2240-1G
TCB2240-25GEA 188 USD
TCB2240-25G TCB2240-5G TCB2240-1G
N-tert-Butylbenzenesulfenamide ≥96.0%
N-tert-Butylbenzenesulfenamide

CAS Number: 19117-31-8
MDL Number: MFCD03844776
Molecular Formula: C10H15NS
Molecular Weight: 181.30
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.01
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfenic Acid Derivatives, Oxidation, Oxidation
Other Product Names / Synonyms: N-tert-Butylphenylsulfenamide

Formula: C₁₀H₁₅NS
Density: 1.01 (20 °C)
MDL Number: MFCD03844776
CAS Number: 19117-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1069ccae8be09d317d25b7813f8a643ab14c21a9&vid=1069ccae8be09d317d25b7813f8a643ab14c21a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR