1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
1-Benzyl-4-hydroxypiperidine-4-carbonitrile hydrochloride
B2237-5G
B2237-25G
TCB2237-5GEA
103
USD
TCB2237-5G
TCB2237-25G
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride ≥98.0%
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride
CAS Number: 71617-20-4
MDL Number: MFCD00012836
Molecular Formula: C13H16N2O
Molecular Weight: 252.74
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 1-Benzyl-4-hydroxypiperidine-4-carbonitrile Hydrochloride
Formula:
C₁₃H₁₇ClN₂O |
MDL Number:
MFCD00012836 CAS Number: 71617-20-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...