(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Bis((R)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine, (+)-2,6-Bis[(4R)-4-(i-propyl)-2-oxazolin-2-yl]pyridine, (R)-(i-Pr)-pybox, (R,R)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)

B2217-1G B2217-250MG B2217-5G
TCB2217-1GEA 189 USD
TCB2217-1G TCB2217-250MG TCB2217-5G
(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0%
(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine

CAS Number: 131864-67-0
MDL Number: MFCD01321334
Molecular Formula: C17H23N3O2
Molecular Weight: 301.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 154
Specific rotation [a]20/D: 118 deg (C=1, CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R,R)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)

Formula: C₁₇H₂₃N₃O₂
MW: 301.39 g/mol
Melting Pt: 154 °C
MDL Number: MFCD01321334
CAS Number: 131864-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b050dc7dfa5d976f525eb8a8e90da84489353ac&vid=0b050dc7dfa5d976f525eb8a8e90da84489353ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR