1-[Bis(2-hydroxyethyl)amino]-2-propanol ≥93.0%

Supplier: TCI America

Synonyms: N,N-Bis(2-hydroxyethyl)isopropanolamine

B2180-25G B2180-5G
TCB2180-25GEA 222 USD
TCB2180-25G TCB2180-5G
1-[Bis(2-hydroxyethyl)amino]-2-propanol ≥93.0%
1-[Bis(2-hydroxyethyl)amino]-2-propanol

CAS Number: 6712-98-7
MDL Number: MFCD00009725
Molecular Formula: C7H17NO3
Molecular Weight: 163.22
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 113


Other Product Names / Synonyms: N,N-Bis(2-hydroxyethyl)isopropanolamine

Formula: C₇H₁₇NO₃
MW: 163.22 g/mol
Boiling Pt: 145 °C
Flash Pt: 113 °C
MDL Number: MFCD00009725
CAS Number: 6712-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36606a95f45116aab2f0f4dbe14e71adbcd100cc&vid=36606a95f45116aab2f0f4dbe14e71adbcd100cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR