2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole ≥98.0%

Supplier: TCI America

B2169-1G B2169-5G
TCB2169-1GEA 71 USD
TCB2169-1G TCB2169-5G
2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole ≥98.0%
2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole

CAS Number: 2425-95-8
MDL Number: MFCD00042667
Molecular Formula: C14H12N4O
Molecular Weight: 252.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: BAO

Formula: C₁₄H₁₂N₄O
MDL Number: MFCD00042667
CAS Number: 2425-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9d84beacf4bc1bd4b01c5b6e23323419395bea9&vid=b9d84beacf4bc1bd4b01c5b6e23323419395bea9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR