(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone ≥96.0%

Supplier: TCI America

Synonyms: (4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone, (S)-4-Benzyl-3-propionyloxazolidin-2-one

B2166-1G B2166-5G
TCB2166-1GEA 44 USD
TCB2166-1G TCB2166-5G
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone ≥96.0%
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone

CAS Number: 101711-78-8 MDL Number: MFCD00269688 Molecular Formula: C13H15NO3 Molecular Weight: 233.27 Purity/Analysis Method: 96.0% (GC) Form: Crystal Melting point (°C): 45 Specific rotation [a]20/D: 103 deg (C=1, EtOH)

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Melting Pt: 45 °C
MDL Number: MFCD00269688
CAS Number: 101711-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=541d28e369fd2db5d654ca4e6c597a3b7de8771e&vid=541d28e369fd2db5d654ca4e6c597a3b7de8771e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR