(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone ≥98.0%

Supplier: TCI America

B2165-5G B2165-1G
TCB2165-5GEA 237 USD
TCB2165-5G TCB2165-1G
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone ≥98.0%
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone

CAS Number: 131685-53-5
MDL Number: MFCD00269687
Molecular Formula: C13H15NO3
Molecular Weight: 233.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45
Specific rotation [a]20/D: -103 deg (C=1, EtOH)

Formula: C₁₃H₁₅NO₃
MW: 233.2631 g/mol
Melting Pt: 45 °C
MDL Number: MFCD00269687
CAS Number: 131685-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44a8c07c65f4dde03840eba33f48910acdd32fb6&vid=44a8c07c65f4dde03840eba33f48910acdd32fb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR