2,2-Bis(3-nitrophenyl)hexafluoropropane ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,3'-(Hexafluoroisopropylidene)bis(nitrobenzene)

B2115-1G B2115-5G
TCB2115-001GEA 139.1 USD
TCB2115-001G TCB2115-5G
2,2-Bis(3-nitrophenyl)hexafluoropropane ≥99.0% (by titrimetric analysis)
2,2-Bis(3-nitrophenyl)hexafluoropropane

CAS Number: 64465-34-5
MDL Number: MFCD02093448
Molecular Formula: C15H8F6N2O4
Molecular Weight: 394.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 120


Other Product Names / Synonyms: 3,3'-(Hexafluoroisopropylidene)bis(nitrobenzene)

Formula: C₁₅H₈F₆N₂O₄
MW: 394.23 g/mol
Melting Pt: 120 °C
MDL Number: MFCD02093448
CAS Number: 64465-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f776472d1826bf741cbc5faa69199883440d3d&vid=d4f776472d1826bf741cbc5faa69199883440d3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR