2-[4-(Bromomethyl)phenyl]propionic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-(Bromomethyl)hydratropic Acid, 2-(4-(Bromomethyl)phenyl)propanoic acid

B2101-5G B2101-25G
TCB2101-005GEA 63.8 USD
TCB2101-005G TCB2101-025G
2-[4-(Bromomethyl)phenyl]propionic acid ≥98.0%
2-[4-(Bromomethyl)phenyl]propionic acid

CAS Number: 111128-12-2
MDL Number: MFCD02093445
Molecular Formula: C10H11BrO2
Molecular Weight: 243.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 4-(Bromomethyl)hydratropic Acid

Formula: C₁₀H₁₁BrO₂
Melting Pt: 125 °C
MDL Number: MFCD02093445
CAS Number: 111128-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2348f921b28a9112a041c7278bb29162c35ad01&vid=e2348f921b28a9112a041c7278bb29162c35ad01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR