4,4'-Bis(chloromethyl)biphenyl ≥95.0%

Supplier: TCI America

Synonyms: 4,4-Bis(chloromethyl)-1,1-biphenyl, 4,4'-Bis(chloromethyl)-1,1-biphenyl

B2099-25G B2099-500G
TCB2099-025GEA 56 USD
TCB2099-025G TCB2099-500G
4,4'-Bis(chloromethyl)biphenyl ≥95.0%
4,4'-Bis(chloromethyl)biphenyl

CAS Number: 1667-10-3
MDL Number: MFCD00674019
Molecular Formula: C14H12Cl2
Molecular Weight: 251.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 184
Melting point (°C): 141

Formula: C₁₄H₁₂Cl₂
MW: 251.15 g/mol
Boiling Pt: 184 °C
Melting Pt: 141 °C
MDL Number: MFCD00674019
CAS Number: 1667-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3cb4da15223e8730b4649020c5b2b442f9bf548&vid=c3cb4da15223e8730b4649020c5b2b442f9bf548&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR