3,5-Dibenzyloxybenzyl bromide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2093-5G B2093-25G
TCB2093-005GEA 277.2 USD
TCB2093-005G TCB2093-25G
3,5-Dibenzyloxybenzyl bromide ≥98.0% (by GC, titration analysis)
3,5-Dibenzyloxybenzyl bromide

CAS Number: 24131-32-6
MDL Number: MFCD02093444
Molecular Formula: C21H19BrO2
Molecular Weight: 383.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 87
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Bis(benzyloxy)benzyl Bromide, 3,5-Dibenzyloxy-alpha-bromotoluene

Formula: C₂₁H₁₉BrO₂
Melting Pt: 87 °C
MDL Number: MFCD02093444
CAS Number: 24131-32-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb71332653ec648e41f86de48ac64dbb438b7927&vid=cb71332653ec648e41f86de48ac64dbb438b7927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR