Boron trifluoride dibutyl etherate (∼30% BF₃)
Supplier: TCI America
|
Danger
|
Synonyms:
Dibutyl ether-boron trifluoride, Boron trifluoride butyl etherate, Boron trifluoride butyl ether, Boron trifluoride-butyl ether complex, 1-butoxybutane- trifluoroborane(1:1), Trifluoroboron dibutyl etherate
B2074-25ML
B2074-500ML
TCB2074-025MLEA
120
USD
TCB2074-025ML
TCB2074-500ML
Boron trifluoride dibutyl etherate (∼30% BF₃)
Boron trifluoride dibutyl etherate
CAS Number: 593-04-4
Molecular Formula: C8H18BF3O
Molecular Weight: 198.04
Form: Liquid
Color: Colorless
Flash Point (°C): 70
Formula:
BF₃·C₈H₁₈O MW: 198.04 g/mol Density: 0.959 Flash Pt: 70°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013195 CAS Number: 593-04-4 UN: 2922 ADR: 8,II |
Specification Test Results
Concentration (Gravimetric Method) | 29.0 to 33.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...