Dipotassium anthraquinone-1,8-disulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Anthraquinone-1,8-disulfonic Acid Dipotassium Salt
, Anthraquinone-1,8-disulphonic Acid Dipotassium Salt
A0505-25G
TCA0505-025GEA
71.03
USD
TCA0505-025G
Dipotassium anthraquinone-1,8-disulfonate ≥98.0% (by titrimetric analysis)
Dipotassium anthraquinone-1,8-disulfonate
CAS Number: 14938-42-2
MDL Number: MFCD00053254
Molecular Formula: C14H8O8S2
Molecular Weight: 444.51
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Anthraquinone-1,8-disulfonic Acid Dipotassium Salt
Formula:
C₁₄H₈O₈S₂ MW: 444.51 g/mol |
MDL Number:
MFCD00053254 CAS Number: 14938-42-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...