2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetrabromobisphenol A Bis(2,3-dibromopropyl) Ether, 2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane(Granular)

B2022-500G B2022-25G
TCB2022-500GEA 54 USD
TCB2022-500G TCB2022-025G
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane ≥95.0% (by titrimetric analysis)
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane

CAS Number: 21850-44-2
MDL Number: MFCD00017887
Molecular Formula: C21H20Br8O2
Molecular Weight: 943.62
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 117


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Tetrabromobisphenol A Bis(2,3-dibromopropyl) Ether

Formula: C₂₁H₂₀Br₈O₂
MW: 943.62 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00017887
CAS Number: 21850-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b00110b65569aef462bf9f6a842d2792fd4f69e&vid=6b00110b65569aef462bf9f6a842d2792fd4f69e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR