4-(2-Bromoethyl)benzenesulfonic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B2006-1G B2006-5G
TCB2006-001GEA 207.1 USD
TCB2006-001G TCB2006-5G
4-(2-Bromoethyl)benzenesulfonic acid ≥97.0% (by titrimetric analysis)
4-(2-Bromoethyl)benzenesulfonic acid

CAS Number: 54322-31-5
MDL Number: MFCD01861279
Molecular Formula: C8H9BrO3S
Molecular Weight: 265.12
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 86

Formula: C₈H₉BrO₃S
MW: 265.12 g/mol
Melting Pt: 86 °C
MDL Number: MFCD01861279
CAS Number: 54322-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e857712d053e339d49e1a7b6ff10c2260e9b761&vid=8e857712d053e339d49e1a7b6ff10c2260e9b761&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR