4-Bromo-m-toluidine ≥98.0%

Supplier: TCI America

Synonyms: 3-Amino-6-bromotoluene, 4-Bromo-3-methylaniline, 5-Amino-2-bromotoluene

B1990-5G B1990-25G
TCB1990-005GEA 17 USD
TCB1990-005G TCB1990-025G
4-Bromo-m-toluidine ≥98.0%
4-Bromo-m-toluidine

CAS Number: 6933-10-4
MDL Number: MFCD00007828
Molecular Formula: C7H8BrN
Molecular Weight: 186.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 240
Melting point (°C): 81


Other Product Names / Synonyms: 5-Amino-2-bromotoluene, 4-Bromo-m-toluidine

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C
Melting Pt: 81 °C
MDL Number: MFCD00007828
CAS Number: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd5d3bfc9057bcdb730836511d0face05853ce3&vid=dfd5d3bfc9057bcdb730836511d0face05853ce3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR