3-(2-Benzothiazolylthio)propionic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-(Benzo[d]thiazol-2-ylthio)propanoic acid
B1942-25G
B1942-500G
TCB1942-025GEA
46
USD
TCB1942-025G
TCB1942-500G
3-(2-Benzothiazolylthio)propionic acid ≥98.0% (by titrimetric analysis)
3-(2-Benzothiazolylthio)propionic acid
CAS Number: 4767-00-4
MDL Number: MFCD00022878
Molecular Formula: C10H9NO2S2
Molecular Weight: 239.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 149
Flash Point (°C): 153
Formula:
C₁₀H₉NO₂S₂ MW: 239.31 g/mol Melting Pt: 149 °C Flash Pt: 153 °C |
MDL Number:
MFCD00022878 CAS Number: 4767-00-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...