Benzenesulfonic anhydride ≥96.0% (by titrimetric analysis)

Supplier: TCI America

B1931-5G B1931-25G
TCB1931-005GEA 180.7 USD
TCB1931-005G TCB1931-25G
Benzenesulfonic anhydride ≥96.0% (by titrimetric analysis)
Benzenesulfonic anhydride

CAS Number: 512-35-6
MDL Number: MFCD00054601
Molecular Formula: C12H10O5S2
Molecular Weight: 298.33
Purity/Analysis Method: >96.0% (T)
Form: Crystal

Formula: C₁₂H₁₀O₅S₂
MW: 298.33 g/mol
MDL Number: MFCD00054601
CAS Number: 512-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4592030f1ca99d66e1a4c12f8c61fc9873417852&vid=4592030f1ca99d66e1a4c12f8c61fc9873417852&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR