9-(Bromomethyl)acridine ≥98.0%

Supplier: TCI America

B1926-1G B1926-5G
TCB1926-001GEA 160.1 USD
TCB1926-001G TCB1926-005G
9-(Bromomethyl)acridine ≥98.0%
9-(Bromomethyl)acridine

[for HPLC Labeling]
CAS Number: 1556-34-9
MDL Number: MFCD01321146
Molecular Formula: C14H10BrN
Molecular Weight: 272.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 165

Formula: C₁₄H₁₀BrN
MW: 272.15 g/mol
Melting Pt: 165 °C
MDL Number: MFCD01321146
CAS Number: 1556-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9caa11c54288a136003420897309996e85e13905&vid=9caa11c54288a136003420897309996e85e13905&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR