4,4'-Bis(bromomethyl)biphenyl ≥95.0%
Supplier: TCI America
Synonyms:
4,4-Bis(bromomethyl)biphenyl
B1921-1G
B1921-5G
TCB1921-001GEA
77
USD
TCB1921-001G
TCB1921-005G
4,4'-Bis(bromomethyl)biphenyl ≥95.0%
4,4'-Bis(bromomethyl)biphenyl
CAS Number: 20248-86-6
MDL Number: MFCD00094419
Molecular Formula: C14H12Br2
Molecular Weight: 340.06
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 171
Formula:
C₁₄H₁₂Br₂ MW: 340.06 g/mol Melting Pt: 171 °C |
MDL Number:
MFCD00094419 CAS Number: 20248-86-6 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...