1,4-Bis(bromomethyl)naphthalene 65.0% (contains isomer)

Supplier: TCI America

B1919-1G B1919-5G
TCB1919-001GEA 183.1 USD
TCB1919-001G TCB1919-5G
1,4-Bis(bromomethyl)naphthalene 65.0% (contains isomer)
1,4-Bis(bromomethyl)naphthalene

CAS Number: 58791-49-4
MDL Number: MFCD01321143
Molecular Formula: C12H10Br2
Molecular Weight: 314.02
Purity/Analysis Method: >65.0% (GC)
Form: Crystal
Melting point (°C): 170

Formula: C₁₂H₁₀Br₂
Melting Pt: 169…173 °C
MDL Number: MFCD01321143
CAS Number: 58791-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=381f37d8d9e805c16172aa5198ddc95c1c5da28e&vid=381f37d8d9e805c16172aa5198ddc95c1c5da28e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >65.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR