4-Biphenyl benzoate ≥98.0%
Supplier: TCI America
Synonyms:
Benzoic Acid 4-Biphenyl Ester, (4-Phenylphenyl) benzoate
B1866-5G
B1866-25G
TCB1866-005GEA
142
USD
TCB1866-005G
TCB1866-025G
4-Biphenyl benzoate ≥98.0%
4-Biphenyl benzoate
CAS Number: 2170-13-0
MDL Number: MFCD00223541
Molecular Formula: C19H14O2
Molecular Weight: 274.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 151
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Benzoic Acid 4-Biphenyl Ester
Formula:
C₁₉H₁₄O₂ MW: 274.32 g/mol Melting Pt: 151 °C |
MDL Number:
MFCD00223541 CAS Number: 2170-13-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...