3-Anilinopropionitrile ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 1075-76-9
MDL Number: MFCD00001953
Molecular Formula: C9H10N2
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 50
Flash Point (°C): 134
Other Product Names / Synonyms: N-(2-Cyanoethyl)aniline
Formula:
C₉H₁₀N₂ Boiling Pt: 160 °C Melting Pt: 50 °C Flash Pt: 134 °C |
MDL Number:
MFCD00001953 CAS Number: 1075-76-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...