3-(2-Methoxyethoxy)propyl bromide ≥95.0%

Supplier: TCI America

Synonyms: 1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether

B1842-1G B1842-5G
TCB1842-001GEA 122.4 USD
TCB1842-001G TCB1842-005G
3-(2-Methoxyethoxy)propyl bromide ≥95.0%
3-(2-Methoxyethoxy)propyl bromide

CAS Number: 59551-75-6
MDL Number: MFCD01321130
Molecular Formula: C6H13BrO2
Molecular Weight: 197.07
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether

Formula: C₆H₁₃BrO₂
MW: 197.07 g/mol
MDL Number: MFCD01321130
CAS Number: 59551-75-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac8a9a654c7a7902545cfdd5ca34657238de805f&vid=ac8a9a654c7a7902545cfdd5ca34657238de805f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR