2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone ≥95.0%

Supplier: TCI America

Synonyms: 2,5-Bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol

B1839-25G B1839-5G
TCB1839-025GEA 88 USD
TCB1839-025G TCB1839-5G
2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone ≥95.0%
2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone

CAS Number: 903-19-5 MDL Number: MFCD00128797 Molecular Formula: C22H38O2 Molecular Weight: 334.54 Purity/Analysis Method: 95.0% (GC) Form: Crystal Melting point (°C): 130

Formula: C₂₂H₃₈O₂
MW: 334.54 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00128797
CAS Number: 903-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43124113dbe432ccfedc91f1b861eec6f4879a58&vid=43124113dbe432ccfedc91f1b861eec6f4879a58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR