2,2'-Bithiophene-5-carboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 2,2-Bithiophene-5-carboxaldehyde, 5-Formyl-2,2'-bithiophene

B1838-25G B1838-1G B1838-5G
TCB1838-25GEA 356 USD
TCB1838-25G TCB1838-001G TCB1838-005G
2,2'-Bithiophene-5-carboxaldehyde ≥98.0%
2,2'-Bithiophene-5-carboxaldehyde

CAS Number: 3779-27-9
MDL Number: MFCD00052294
Molecular Formula: C9H6OS2
Molecular Weight: 194.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 57


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 5-Formyl-2,2'-bithiophene

Formula: C₉H₆OS₂
MW: 194.27 g/mol
Boiling Pt: 210 °C
Melting Pt: 57 °C
MDL Number: MFCD00052294
CAS Number: 3779-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2ec317269a13c5002762d5e3d58bfc8b2713ba6&vid=d2ec317269a13c5002762d5e3d58bfc8b2713ba6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR