β-methasone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9ɑ-Fluoro-11β,17ɑ,21-trihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione

B1837-1G B1837-5G
TCB1837-001GEA 74.27 USD
TCB1837-001G TCB1837-005G
β-methasone ≥98.0% (by HPLC)
Betamethasone

CAS Number: 378-44-9
MDL Number: MFCD00062969
Molecular Formula: C22H29FO5
Molecular Weight: 392.47
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 118 deg (C=1, Dioxane)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Pregnanes (Glucocorticoi
Other Product Names / Synonyms: 9alpha-Fluoro-11beta,17alpha,21-trihydroxy-16beta-methyl-1,4-pregnadiene-3,20-dione

Formula: C₂₂H₂₉FO₅
MW: 392.47 g/mol
MDL Number: MFCD00062969
CAS Number: 378-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ed4f6308f8d9fa01295eb7cdfa07f8b90e34bf3&vid=6ed4f6308f8d9fa01295eb7cdfa07f8b90e34bf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR