(1-Bromovinyl)trimethylsilane ≥96.0%
Supplier: TCI America
Synonyms:
1-Bromo-1-trimethylsilylethylene, alpha-Trimethylsilylvinyl Bromide
B1769-5G
B1769-1G
TCB1769-005GEA
196
USD
TCB1769-005G
TCB1769-1G
(1-Bromovinyl)trimethylsilane ≥96.0%
(1-Bromovinyl)trimethylsilane
CAS Number: 13683-41-5
MDL Number: MFCD00014927
Molecular Formula: C5H11BrSi
Molecular Weight: 179.13
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): 9
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: 1-Bromo-1-trimethylsilylethylene, alpha-Trimethylsilylvinyl Bromide
Formula:
C₅H₁₁BrSi MW: 179.13 g/mol Boiling Pt: 128 °C Density: 1.16 (20 °C) Flash Pt: 9 °C |
MDL Number:
MFCD00014927 CAS Number: 13683-41-5 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...