1,3-Bis(4-methoxyphenyl)-1,3-propanedione ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Bis(4-methoxyphenyl)propane-1,3-dione

B1733-25G B1733-5G
TCB1733-25GEA 539 USD
TCB1733-25G TCB1733-005G
1,3-Bis(4-methoxyphenyl)-1,3-propanedione ≥98.0%
1,3-Bis(4-methoxyphenyl)-1,3-propanedione

CAS Number: 18362-51-1
MDL Number: MFCD00025817
Molecular Formula: C17H16O4
Molecular Weight: 284.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 116

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00025817
CAS Number: 18362-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11cec059a0356add8d3b3ff66334b290b180f8f6&vid=11cec059a0356add8d3b3ff66334b290b180f8f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR