9,9-Bis(4-hydroxyphenyl)fluorene ≥96.0%
Supplier: TCI America
CAS Number: 3236-71-3
MDL Number: MFCD00191392
Molecular Formula: C25H18O2
Molecular Weight: 350.42
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 227
Flash Point (°C): 200
Product Categories: Materials Chemistry, Monomers, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4,4'-(9-Fluorenylidene)diphenol
Formula:
C₂₅H₁₈O₂ Melting Pt: 227 °C Flash Pt: 200 °C |
MDL Number:
MFCD00191392 CAS Number: 3236-71-3 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...