(2-Bromoallyl)trimethylsilane ≥93.0%

Supplier: TCI America

B1695-1G B1695-5G
TCB1695-001GEA 286.2 USD
TCB1695-001G TCB1695-5G
(2-Bromoallyl)trimethylsilane ≥93.0%
(2-Bromoallyl)trimethylsilane

CAS Number: 81790-10-5
MDL Number: MFCD00043054
Molecular Formula: C6H13BrSi
Molecular Weight: 193.16
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Flash Point (°C): 30
Specific Gravity (20/20): 1.13
Storage Temperature: <0°C

Formula: C₆H₁₃BrSi
MW: 193.16 g/mol
Density: 1.13 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00043054
CAS Number: 81790-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52aec7f70d3c4b9e822fe0f00bbabfdc8f5ae938&vid=52aec7f70d3c4b9e822fe0f00bbabfdc8f5ae938&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR