Pentyl nitrite (mixed isomers ) ≥95.0%

Supplier: TCI America

A0455-500ML A0455-25ML A0455-100ML
TCA0455-500MLEA 651 USD
TCA0455-500ML TCA0455-025ML TCA0455-100ML
Pentyl nitrite (mixed isomers ) ≥95.0%
Pentyl nitrite (mixed isomers )

CAS Number: 463-04-7
MDL Number: MFCD00036132
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): -18
Specific Gravity (20/20): 0.88
Storage Temperature: 0-10°C


Product Categories: Bioscience, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Pentyl Nitrite

Formula: C₅H₁₁NO₂
Boiling Pt: 104 °C
Density: 0.88 (20 °C)
Flash Pt: –18 °C
MDL Number: MFCD00036132
CAS Number: 463-04-7
EINECS: 207-332-7
UN: 1113
ADR: 3,II
Merck Index: 12,01617
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc6b7235eb9643ea0941bfdd8bd4fd90434297ec&vid=bc6b7235eb9643ea0941bfdd8bd4fd90434297ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR