N-(tert-Butoxycarbonyl)-p-toluenesulfonamide ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-p-toluenesulphonamide, tert-Butyl tosylcarbamate

B1648-25G B1648-10G
TCB1648-25GEA 130 USD
TCB1648-25G TCB1648-010G
N-(tert-Butoxycarbonyl)-p-toluenesulfonamide ≥98.0%
N-(tert-Butoxycarbonyl)-p-toluenesulfonamide

CAS Number: 18303-04-3
MDL Number: MFCD00134267
Molecular Formula: C12H17NO4S
Molecular Weight: 271.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 123


Product Categories: Synthetic Organic Chemistry, Amination
Other Product Names / Synonyms: N-Boc-p-toluenesulfonamide

Formula: C₁₂H₁₇NO₄S
Melting Pt: 123 °C
MDL Number: MFCD00134267
CAS Number: 18303-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d85294080fcb4b89b5dae32fb6114203d319635&vid=4d85294080fcb4b89b5dae32fb6114203d319635&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR