5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Benzyl Ester, N-Boc-L-glutamic Acid 5-Benzyl Ester, 5-Benzyl N-Boc-L-glutamate, Boc-Glu(OBzl)-OH

B1633-25G B1633-5G
TCB1633-25GEA 129 USD
TCB1633-25G TCB1633-005G
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate ≥98.0% (by HPLC, titration analysis)
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate

CAS Number: 13574-13-5
MDL Number: MFCD00065569
Molecular Formula: C17H23NO6
Molecular Weight: 337.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 68
Specific rotation [a]20/D: -16.5 deg (C=2, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-Benzyl N-Boc-L-glutamate, N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Benzyl Ester, N-Boc-L-glutamic Acid 5-Benzyl Ester, Boc-Glu(OBzl)-OH

Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00065569
CAS Number: 13574-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15f3365e31d9724cb81e7dc7ae7dd634e2b2b93d&vid=15f3365e31d9724cb81e7dc7ae7dd634e2b2b93d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR