Dipentyl ether ≥98.0%

Supplier: TCI America

Synonyms: Amyl ether, Di-n-amyl ether, n-Pentyl ether

A0451-25ML A0451-500ML
TCA0451-025MLEA 63 USD
TCA0451-025ML TCA0451-500ML
Dipentyl ether ≥98.0%
Dipentyl ether

CAS Number: 693-65-2 MDL Number: MFCD00009501 Molecular Formula: C10H22O Molecular Weight: 158.29 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 186 Melting point (°C): -69 Flash Point (°C): 57 Specific Gravity (20/20): 0.78


Form: Clear Liquid


Other Product Names / Synonyms: Diamyl Ether, Pentyl Ether, Dipentyl Ether

Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 186 °C
Melting Pt: –69 °C
Density: 0.78 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00009501
CAS Number: 693-65-2
EINECS: 211-756-8
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54e09e899322fc43f99ea9b9175ffc8ec88d5db9&vid=54e09e899322fc43f99ea9b9175ffc8ec88d5db9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR