7-Bromobenz[a]anthracene ≥98.0%
Supplier: TCI America
B1603-5G
B1603-1G
TCB1603-5GEA
571.2
USD
TCB1603-5G
TCB1603-001G
7-Bromobenz[a]anthracene ≥98.0%
7-Bromobenz[a]anthracene
CAS Number: 32795-84-9
MDL Number: MFCD00029016
Molecular Formula: C18H11Br
Molecular Weight: 307.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152
Formula:
C₁₈H₁₁Br Melting Pt: 152 °C |
MDL Number:
MFCD00029016 CAS Number: 32795-84-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...