(3-Bromopropyl)phosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Bromopropanephosphonic Acid, (3-Bromopropyl)phosphonicacid

B1591-1G B1591-5G
TCB1591-001GEA 79 USD
TCB1591-001G TCB1591-5G
(3-Bromopropyl)phosphonic acid ≥98.0% (by titrimetric analysis)
(3-Bromopropyl)phosphonic acid

CAS Number: 1190-09-6
MDL Number: MFCD00191385
Molecular Formula: C3H8BrO3P
Molecular Weight: 202.97
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 112


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 3-Bromopropanephosphonic Acid

Formula: C₃H₈BrO₃P
MW: 202.97 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00191385
CAS Number: 1190-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1935b9bfc201d5d3f4127d253076128f16e61fa9&vid=1935b9bfc201d5d3f4127d253076128f16e61fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR