(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine ≥98.0%

Supplier: TCI America

B1583-25G B1583-5G B1583-1G
TCB1583-25GEA 726 USD
TCB1583-25G TCB1583-005G TCB1583-1G
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine ≥98.0%
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine

CAS Number: 169749-99-9
MDL Number: MFCD00191310
Molecular Formula: C12H18N2
Molecular Weight: 190.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.99
Specific rotation [a]20/D: 3 deg (C=10, EtOH)

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Density: 0.99 (20 °C)
MDL Number: MFCD00191310
CAS Number: 169749-99-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48574324faf0f1f7acb5f1591031148962d6f37e&vid=48574324faf0f1f7acb5f1591031148962d6f37e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR