(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine ≥95.0%
Supplier: TCI America
B1581-5G
B1581-1G
TCB1581-005GEA
896
USD
TCB1581-005G
TCB1581-1G
(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine ≥95.0%
(3S)-(+)-1-Benzyl-3-(ethylamino)pyrrolidine
CAS Number: 169750-99-6
MDL Number: MFCD00191307
Molecular Formula: C13H20N2
Molecular Weight: 204.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: 8 deg (C=10, EtOH)
Formula:
C₁₃H₂₀N₂ Density: 0.98 (20 °C) |
MDL Number:
MFCD00191307 CAS Number: 169750-99-6 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...