1,1-Bis(4-hydroxyphenyl)cyclohexane ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Cyclohexylidenebisphenol
B1568-25G
B1568-500G
B1568-100G
TCB1568-025GEA
50
USD
TCB1568-025G
TCB1568-500G
TCB1568-100G
1,1-Bis(4-hydroxyphenyl)cyclohexane ≥98.0%
1,1-Bis(4-hydroxyphenyl)cyclohexane
CAS Number: 843-55-0
MDL Number: MFCD00019341
Molecular Formula: C18H20O2
Molecular Weight: 268.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 189
Flash Point (°C): 246
Product Categories: Materials Chemistry, Monomers, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4,4'-Cyclohexylidenebisphenol
Formula:
C₁₈H₂₀O₂ MW: 268.36 g/mol Melting Pt: 189 °C Flash Pt: 246 °C |
MDL Number:
MFCD00019341 CAS Number: 843-55-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...