Pentylamine mixture of isomers ≥98.0%

Supplier: TCI America

Synonyms: Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine

A0446-25ML A0446-500ML
TCA0446-025MLEA 28 USD
TCA0446-025ML TCA0446-500ML
Pentylamine mixture of isomers ≥98.0%
Pentylamine

MDL Number: MFCD00008236
Molecular Formula: C5H13N
Molecular Weight: 87.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): -1
Specific Gravity (20/20): 0.76

Formula: H₃C(CH₂)₃CH₂NH₂
MW: 87.16 g/mol
Density: 0.76 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD00008236
CAS Number: 110-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79b7c3b68d8b3fbed377337bbc551c4b859f35c1&vid=748b1584dabec1f60b6f6a3713c868191d71fb75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR