1,3-Bis(isocyanatomethyl)cyclohexane (cis and trans mixture) ≥99.0%

Supplier: TCI America

B1538-500ML B1538-25ML
TCB1538-500MLEA 161.74 USD
TCB1538-500ML TCB1538-025ML
1,3-Bis(isocyanatomethyl)cyclohexane (cis and trans mixture) ≥99.0%
1,3-Bis(isocyanatomethyl)cyclohexane (cis and trans mixture)

CAS Number: 38661-72-2
MDL Number: MFCD00129947
Molecular Formula: C10H14N2O2
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Flash Point (°C): 159
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Density: 1.10 (20 °C)
Flash Pt: 159 °C
MDL Number: MFCD00129947
CAS Number: 38661-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9be464dff2b642062e1a43988335d7b6109ac41&vid=e9be464dff2b642062e1a43988335d7b6109ac41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR