2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

B1535-1G B1535-100MG
TCB1535-1GEA 710 USD
TCB1535-1G TCB1535-100MG
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide ≥97.0% (by HPLC, titration analysis)
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide

CAS Number: 220191-36-6 MDL Number: MFCD00142790 Molecular Formula: C18H19NO2S2 Molecular Weight: 345.48 Purity/Analysis Method: 97.0% (HPLC,T) Form: Crystal Melting point (°C): 181

Formula: C₁₈H₁₉NO₂S₂
MW: 345.48 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00142790
CAS Number: 220191-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b9d2a075aad80a43a1a38f79ab62384670c6304&vid=2b9d2a075aad80a43a1a38f79ab62384670c6304&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR