1,1'-Bis(2,4-dinitrophenyl)-4,4'-bipyridinium dichloride ≥97.0% (by HPLC)

Supplier: TCI America

B1518-25G B1518-5G
TCB1518-025GEA 222 USD
TCB1518-025G TCB1518-005G
1,1'-Bis(2,4-dinitrophenyl)-4,4'-bipyridinium dichloride ≥97.0% (by HPLC)
1,1'-Bis(2,4-dinitrophenyl)-4,4'-bipyridinium dichloride

CAS Number: 41168-79-0
MDL Number: MFCD00142352
Molecular Formula: C22H14Cl2N6O8
Molecular Weight: 561.29
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal

Formula: C₂₂H₁₄Cl₂N₆O₈
MW: 561.29 g/mol
MDL Number: MFCD00142352
CAS Number: 41168-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83494f7e30a591cb71323f57f994a9e19b2e78dc&vid=83494f7e30a591cb71323f57f994a9e19b2e78dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR