3-Butylthiophene ≥98.0%

Supplier: TCI America

B1458-1G B1458-5G
TCB1458-001GEA 26 USD
TCB1458-001G TCB1458-005G
3-Butylthiophene ≥98.0%
3-Butylthiophene

CAS Number: 34722-01-5 MDL Number: MFCD00143179 Molecular Formula: C8H12S Molecular Weight: 140.24 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 183 Flash Point (°C): 60 Specific Gravity (20/20): 0.96

Formula: C₈H₁₂S
MW: 140.2459 g/mol
Boiling Pt: 183 °C
Density: 0.96 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00143179
CAS Number: 34722-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=420400c351b83d42330b0056ce3603434dcac8fd&vid=420400c351b83d42330b0056ce3603434dcac8fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR