O-Benzyl-L-serine methyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
H-Ser(Bzl)-OMe.HCl, H-Ser(Bzl)-OMe·HCl, Methyl O-Benzyl-L-serinate Hydrochloride
B1450-1G
B1450-5G
TCB1450-001GEA
90
USD
TCB1450-001G
TCB1450-005G
O-Benzyl-L-serine methyl ester hydrochloride ≥98.0%
O-Benzyl-L-serine methyl ester hydrochloride
CAS Number: 19525-87-2
MDL Number: MFCD00191312
Molecular Formula: C11H15NO3
Molecular Weight: 245.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 5 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Methyl O-Benzyl-L-serinate Hydrochloride, H-Ser(Bzl)-OMe.HCl
Formula:
C₁₁H₁₅NO₃ MW: 245.70 g/mol |
MDL Number:
MFCD00191312 CAS Number: 19525-87-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...