N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97.0%

Supplier: TCI America

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

B1445-5G B1445-1G
TCB1445-005GEA 302 USD
TCB1445-005G TCB1445-1G
N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97.0%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine

CAS Number: 4741-99-5
MDL Number: MFCD00008174
Molecular Formula: C7H20N4
Molecular Weight: 160.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 287
Flash Point (°C): 110
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers, Bioscience, Polyamines
Other Product Names / Synonyms: 3,7-Diaza-1,9-nonanediamine

Formula: C₇H₂₀N₄
MW: 160.27 g/mol
Boiling Pt: 287 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008174
CAS Number: 4741-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bce14eb64e0832fb6e65654c488b6ba8d9bcb2de&vid=bce14eb64e0832fb6e65654c488b6ba8d9bcb2de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR