Sodium-4-aminosalicylate dihydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A0423-500G A0423-25G
TCA0423-500GEA 370 USD
TCA0423-500G TCA0423-025G
Sodium-4-aminosalicylate dihydrate ≥98.0% (by HPLC, total nitrogen)
Sodium 4-aminosalicylate dihydrate

CAS Number: 6018-19-5
MDL Number: MFCD00064392
Molecular Formula: C7H7NO3
Molecular Weight: 175.12
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: 4-Amino-2-hydroxybenzoic Acid Sodium Salt, 4-Aminosalicylic Acid Sodium Salt

Formula: C₇H₆NNaO₃·2H₂O
MDL Number: MFCD00064392
CAS Number: 6018-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=076b83035ff4de28cfc79c01fb598efb0b0d313f&vid=076b83035ff4de28cfc79c01fb598efb0b0d313f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR