Benzylhydrazine monohydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Benzylhydrazine hydrochloride

B1396-5G B1396-25G
TCB1396-005GEA 113 USD
TCB1396-005G TCB1396-25G
Benzylhydrazine monohydrochloride ≥98.0% (by HPLC)
Benzylhydrazine monohydrochloride

CAS Number: 1073-62-7
MDL Number: MFCD00012921
Molecular Formula: C7H10N2
Molecular Weight: 158.63
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 113

Formula: C₇H₁₀N₂
MW: 158.63 g/mol
Melting Pt: 113 °C
MDL Number: MFCD01722685
CAS Number: 1073-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be5a15a8b2a0e3d2cc8a1fdfaa4009ea067dd93f&vid=be5a15a8b2a0e3d2cc8a1fdfaa4009ea067dd93f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR