1-Benzyl-3-acetamidopyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Acetamido-1-benzylpyrrolidine

B1355-25G B1355-5G
TCB1355-025GEA 116.1 USD
TCB1355-025G TCB1355-5G
1-Benzyl-3-acetamidopyrrolidine ≥98.0% (by GC, titration analysis)
1-Benzyl-3-acetamidopyrrolidine

CAS Number: 28506-01-6
MDL Number: MFCD00059037
Molecular Formula: C13H18N2O
Molecular Weight: 218.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 91


Other Product Names / Synonyms: 3-Acetamido-1-benzylpyrrolidine

Formula: C₁₃H₁₈N₂O
MW: 218.30 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00059037
CAS Number: 28506-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd548e1105a4f46cacade81fdf5d2f031ef3c484&vid=dd548e1105a4f46cacade81fdf5d2f031ef3c484&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR